Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipsnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008698.2 | NP_032724.1 | 1, 2 | 104 | ||
GenBank mRNA | AJ001260.1 | - | 1, 2 | 104 | ||
AK013140.1 | - | 1, 2 | 112 | |||
AK031804.1 | - | 1, 2 | 120 | |||
BC010837.1 | - | 1, 2 | 102 | 4242309 |