Search Thermo Fisher Scientific
Gene Symbol: Nipsnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008698.2 | NP_032724.1 | 3 | 241 | ||
GenBank mRNA | AJ001260.1 | - | 3 | 241 | ||
AK013140.1 | - | 3 | 249 | |||
AK031804.1 | - | 3 | 257 | |||
AK147786.1 | - | 4 | 518 | |||
BC010837.1 | - | 3 | 239 | 4242309 |