Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053074.1 | NP_444304.1 | 2 | 1706 | ||
GenBank mRNA | AK147894.1 | - | 2 | 1736 | ||
AK149637.1 | - | 2 | 1757 | |||
AK150087.1 | - | 2 | 1727 | |||
AK155059.1 | - | 2 | 1731 | |||
AK166621.1 | - | 2 | 1874 | |||
AK168166.1 | - | 2 | 1728 | |||
AK168486.1 | - | 2 | 1739 | |||
AK169178.1 | - | 2 | 1750 | |||
AK171455.1 | - | 2 | 2041 | |||
AK171611.1 | - | 2 | 2039 | |||
AK216353.1 | - | 1 | 30 | |||
BC005784.1 | - | 2 | 1706 | 3603287 | ||
BC089317.1 | - | 2 | 1787 | 6829463 | ||
S59342.1 | - | 2 | 1704 |