Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008776.2 | NP_032802.1 | 6 | 702 | ||
GenBank mRNA | AK154547.1 | - | 6 | 698 | ||
AK167496.1 | - | 6 | 670 | |||
BC067015.1 | - | 6 | 673 | 6820858 | ||
U57746.1 | - | 6 | 700 |