Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008894.2 | NP_032920.2 | 8 | 1021 | ||
GenBank mRNA | AK013901.1 | - | 8 | 987 | ||
BC003342.1 | - | 8 | 987 | 3257610 | ||
CB205181.1 | - | 5 | 557 | |||
Z72486.1 | - | 8 | 982 |