Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019424.2 | NP_062297.1 | 16, 17 | 1812 | ||
GenBank mRNA | AK031397.1 | - | 16, 17 | 1826 | ||
AK053974.1 | - | 14, 15 | 1645 | |||
AK154994.1 | - | 16, 17 | 2001 | |||
AK171225.1 | - | 16, 17 | 1788 | |||
BC046329.1 | - | 14, 15 | 1633 | |||
BC058332.1 | - | 16, 17 | 1795 | |||
BC089331.1 | - | 16, 17 | 1795 | 6848298 | ||
U78315.1 | - | 15, 16 | 1747 | |||
U97149.1 | - | 15, 16 | 1761 |