Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019424.2 | NP_062297.1 | 13 | 1504 | ||
GenBank mRNA | AK031397.1 | - | 13 | 1518 | ||
AK053974.1 | - | 11 | 1337 | |||
AK154994.1 | - | 13 | 1693 | |||
AK171225.1 | - | 13 | 1480 | |||
AK181595.1 | - | 1 | 47 | |||
AK211068.1 | - | 1 | 47 | |||
BC046329.1 | - | 11 | 1325 | |||
BC058332.1 | - | 13 | 1487 | |||
BC089331.1 | - | 13 | 1487 | 6848298 | ||
U78315.1 | - | 12 | 1439 | |||
U97149.1 | - | 12 | 1453 |