Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159730.1 | NP_001153202.1 | 4 | 667 | ||
NM_024458.2 | NP_077778.1 | 4 | 655 | |||
GenBank mRNA | BC012507.1 | - | 4 | 641 | 4500900 | |
BC021920.1 | - | 4 | 656 | 4506350 | ||
BC037176.1 | - | 3 | 2770 | |||
L08075.1 | - | 4 | 674 |