Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009160.2 | NP_033186.1 | 8 | 1011 | ||
GenBank mRNA | AI573415.1 | - | 1 | 314 | ||
BC003705.1 | - | 8 | 962 | 3601620 | ||
BG085186.2 | - | 4 | 450 | |||
CJ237327.1 | - | 1 | 195 | |||
L40156.1 | - | 7 | 939 |