Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144786.2 | NP_659035.2 | 14, 15 | 1871 | ||
GenBank mRNA | AF332053.1 | - | 2, 3 | 169 | ||
AF332054.1 | - | 2, 3 | 169 | |||
AK032051.1 | - | 14, 15 | 1869 | |||
AK131175.1 | - | 11, 12 | 3894 | |||
BC005772.1 | - | 14, 15 | 1852 | 3602314 |