Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bzrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172449.2 | NP_766037.2 | 24 | 5784 | ||
GenBank mRNA | AK051895.1 | - | 3 | 1733 | ||
AK122329.1 | - | 2 | 655 | |||
BC055691.1 | - | 4 | 535 | |||
BC066146.1 | - | 3 | 795 | 30470084 | ||
BC083181.1 | - | 6 | 758 |