Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040398.2 | NP_001035488.2 | 15 | 5829 | ||
GenBank mRNA | AK122435.1 | - | 7 | 2164 | ||
AK140234.1 | - | 1 | 690 | |||
BC035291.1 | - | 7 | 2331 | |||
BC038367.1 | - | 9 | 2907 | |||
BC040775.1 | - | 7 | 2331 | |||
BC041681.1 | - | 7 | 2350 |