Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114116.1 | NP_001107588.1 | 5, 6 | 1269 | ||
NM_016663.3 | NP_057872.3 | 5, 6 | 1296 | |||
GenBank mRNA | AB000893.1 | - | 3, 4 | 1059 | ||
BC051969.1 | - | 4, 5 | 1177 | 5694653 | ||
D45858.1 | - | 4, 5 | 1230 |