Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033249.3 | NP_001028421.1 | 5 | 1678 | ||
GenBank mRNA | AK133380.1 | - | 5 | 1680 | ||
BC056221.1 | - | 3 | 1267 |