Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153143.4 | NP_694783.1 | 1 | 1046 | ||
GenBank mRNA | AF465352.1 | - | 1 | 1013 | ||
AK039558.1 | - | 1468 | ||||
AK161602.1 | - | 1 | 1041 | |||
AK170258.1 | - | 1 | 940 | |||
BC052502.1 | - | 1 | 294 | 5119264 | ||
BC052734.1 | - | 1 | 997 | 6816736 | ||
BC063052.1 | - | 1 | 997 |