Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178680.4 | NP_848795.3 | 8 | 1070 | ||
GenBank mRNA | AF539793.1 | - | 7 | 963 | ||
AK052294.1 | - | 8 | 997 | |||
AK081821.1 | - | 8 | 1009 | |||
AK084418.1 | - | 8 | 1038 | |||
AK168968.1 | - | 8 | 1039 | |||
AK169274.1 | - | 8 | 1009 | |||
BC084585.1 | - | 8 | 1003 | 30429260 |