Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178680.4 | NP_848795.3 | 7 | 775 | ||
GenBank mRNA | AF539793.1 | - | 6 | 668 | ||
AK052294.1 | - | 7 | 702 | |||
AK081821.1 | - | 7 | 714 | |||
AK084418.1 | - | 7 | 743 | |||
AK168968.1 | - | 7 | 744 | |||
AK169274.1 | - | 7 | 714 | |||
BC084585.1 | - | 7 | 708 | 30429260 |