Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nags | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145829.2 | NP_665828.1 | 4 | 1125 | ||
GenBank mRNA | AF462069.1 | - | 4 | 1081 | ||
AJ489814.1 | - | 4 | 1051 | |||
AK075765.1 | - | 3 | 771 | |||
BC036161.1 | - | 2 | 1511 | |||
BC057990.1 | - | 4 | 1089 | 5100021 |