Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126330.1 | NP_001119802.1 | 4, 5 | 831 | ||
NM_001126331.1 | NP_001119803.1 | 4, 5 | 831 | |||
NM_011642.3 | NP_035772.2 | 6, 7 | 1121 | |||
GenBank mRNA | AB221594.1 | - | 5, 6 | 713 | ||
AK017412.1 | - | 3, 4 | 362 | |||
AK139633.1 | - | 4, 5 | 832 | |||
BC066045.1 | - | 4, 5 | 718 | 6812399 | ||
Y19234.1 | - | 6, 7 | 1122 | |||
Y19235.1 | - | 4, 5 | 825 |