Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009478.3 | NP_033504.2 | 7, 8 | 803 | ||
GenBank mRNA | AK002844.1 | - | 7, 8 | 793 | ||
AK172426.1 | - | 7, 8 | 814 | |||
AK211828.1 | - | 2, 3 | 306 | |||
BC008109.1 | - | 7, 8 | 803 | 3481699 |