Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C030006K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145472.2 | NP_663447.1 | 2 | 225 | ||
NM_176828.4 | NP_789798.2 | 2 | 287 | |||
GenBank mRNA | AK053546.1 | - | 2 | 203 | ||
AK075645.1 | - | 2 | 221 | |||
AK085641.1 | - | 2 | 499 | |||
AK140368.1 | - | 1 | 656 | |||
AK140578.1 | - | 2 | 288 | |||
AK163615.1 | - | 2 | 208 | |||
AK165937.1 | - | 2 | 219 | |||
AK186599.1 | - | 2 | 132 | |||
BC019462.1 | - | 2 | 201 | 4458439 | ||
BC026997.1 | - | 2 | 199 | 5362226 |