Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E330017A01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175011.2 | NP_778176.1 | 4 | 463 | ||
GenBank mRNA | AK087761.1 | - | 4 | 464 | ||
BC099383.1 | - | 4 | 499 | 30936622 |