Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trerf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097623.1 | NP_001091092.1 | 13 | 3242 | ||
NM_172622.2 | NP_766210.1 | 10 | 3019 | |||
GenBank mRNA | AK035176.1 | - | 8 | 2791 | ||
AK046822.1 | - | 8 | 2660 | |||
AK154542.1 | - | 11 | 2929 | |||
AK155404.1 | - | 11 | 2926 | |||
AY081774.1 | - | 9 | 2929 | |||
BC059215.1 | - | 13 | 3242 | 6412734 | ||
BC108416.1 | - | 8 | 2702 | 5697521 |