Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146093.1 | NP_666205.1 | 3, 4 | 169 | ||
GenBank mRNA | AK015468.1 | - | 2, 3 | 221 | ||
AK150442.1 | - | 2, 3 | 172 | |||
AK159708.1 | - | 2, 3 | 145 | |||
AK172428.1 | - | 1, 2 | 420 | |||
BC006701.1 | - | 3, 4 | 169 | 3583916 |