Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146093.1 | NP_666205.1 | 7, 8 | 595 | ||
GenBank mRNA | AK150442.1 | - | 6, 7 | 598 | ||
AK159708.1 | - | 6, 7 | 571 | |||
AK185583.1 | - | 2, 3 | 102 | |||
BC006701.1 | - | 7, 8 | 595 | 3583916 |