Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289765.1 | NP_001276694.1 | 5 | 924 | ||
NM_001289766.1 | NP_001276695.1 | 5 | 889 | |||
NM_009539.3 | NP_033565.2 | 5 | 881 | |||
GenBank mRNA | AK039883.1 | - | 5 | 818 | ||
AK089056.1 | - | 5 | 854 | |||
BC029727.1 | - | 5 | 839 | 4925739 | ||
U04379.1 | - | 5 | 835 | |||
U77667.1 | - | 4 | 727 |