Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172748.2 | NP_766336.2 | 9 | 1948 | ||
GenBank mRNA | AK028405.1 | - | 9 | 1715 | ||
AK031431.1 | - | 7 | 1341 | |||
AK162161.1 | - | 3 | 327 | |||
BC053109.1 | - | 5 | 941 | |||
BC059812.1 | - | 9 | 1948 | 6401846 |