Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045553.1 | NP_001039018.1 | 5 | 761 | ||
NM_172754.2 | NP_766342.2 | 5 | 776 | |||
GenBank mRNA | AK083009.1 | - | 5 | 743 | ||
AK139223.1 | - | 5 | 762 | |||
AK168306.1 | - | 5 | 766 | |||
BC010277.1 | - | 4 | 380 | 3586128 |