Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fez1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183171.4 | NP_898994.2 | 1 | 136 | ||
GenBank mRNA | AK138656.1 | - | 2 | 361 | ||
BC029629.1 | - | 2 | 241 | 4035444 | ||
DQ141674.1 | - | 1 | 140 |