Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206924.2 | NP_996807.1 | 5 | 720 | ||
GenBank mRNA | AB016490.1 | - | 5 | 665 | ||
AK004469.1 | - | 5 | 632 | |||
AK089048.1 | - | 5 | 632 | |||
AK215601.1 | - | 3 | 234 | |||
BC008139.1 | - | 5 | 684 | 3582930 | ||
BY617278.1 | - | 1 | 32 |