Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015751.2 | NP_056566.2 | 12, 13 | 1439 | ||
GenBank mRNA | AK049299.1 | - | 12, 13 | 1441 | ||
AK133748.1 | - | 12, 13 | 1370 | |||
AK134738.1 | - | 12, 13 | 1376 | |||
AK144711.1 | - | 12, 13 | 1371 | |||
AK147151.1 | - | 12, 13 | 1377 | |||
AK167130.1 | - | 12, 13 | 1387 | |||
AK167370.1 | - | 12, 13 | 1377 | |||
AK167704.1 | - | 12, 13 | 1338 | |||
BC005422.1 | - | 12, 13 | 1360 | 3489529 | ||
BC066794.1 | - | 11, 12 | 1142 | |||
BC066836.1 | - | 11, 12 | 1142 | |||
U90446.1 | - | 12, 13 | 1328 |