Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172845.2 | NP_766433.1 | 7 | 2201 | ||
GenBank mRNA | AK028364.1 | - | 2202 | |||
AK076295.1 | - | 6 | 1576 | |||
AK083534.1 | - | 7 | 2201 | |||
AK156429.1 | - | 5 | 784 | |||
AK157287.1 | - | 2202 | ||||
AK164900.1 | - | 7 | 2197 | |||
AK173001.1 | - | 2079 | ||||
BC027773.1 | - | 7 | 1835 | 5345809 |