Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011906.2 | NP_036036.2 | 11 | 1227 | ||
GenBank mRNA | AF051098.1 | - | 11 | 1034 | ||
AK089317.1 | - | 11 | 1098 | |||
AK164913.1 | - | 11 | 984 | |||
AK172045.1 | - | 11 | 1158 | |||
AK182265.1 | - | 2 | 124 | |||
AK206088.1 | - | 2 | 124 | |||
BC010244.1 | - | 9 | 783 | |||
BC028977.1 | - | 11 | 1141 | 5363085 | ||
BC057296.1 | - | 11 | 1160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC012634.1 | - | 606 | Chr. - - [-] on Build |