Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175525.3 | NP_780734.1 | 5 | 411 | ||
GenBank mRNA | AK085578.1 | - | 5 | 412 | ||
BC116438.2 | - | 5 | 407 | 40104565 | ||
BC116439.2 | - | 5 | 407 | 40104566 |