Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172920.4 | NP_766508.3 | 21 | 2027 | ||
GenBank mRNA | AK031291.1 | - | 3 | 1107 | ||
AK143845.1 | - | 22 | 2032 | |||
BC031382.1 | - | 14 | 1205 | |||
BC116782.1 | - | 3 | 595 | |||
BC116784.1 | - | 3 | 595 |