Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168508.1 | NP_001161980.1 | 6 | 866 | ||
NM_001168513.1 | NP_001161985.1 | 6 | 467 | |||
NM_001168514.1 | NP_001161986.1 | 7 | 911 | |||
NM_011951.3 | NP_036081.1 | 6 | 866 | |||
GenBank mRNA | AF128892.1 | - | 6 | 764 | ||
AK089059.1 | - | 6 | 780 | |||
AK133684.1 | - | 6 | 779 | |||
AK151348.1 | - | 6 | 468 | |||
AK153025.1 | - | 6 | 468 | |||
AK199780.1 | - | 208 | ||||
BC012235.1 | - | 6 | 866 | 4195842 | ||
BC018227.1 | - | 7 | 808 | |||
BC085303.1 | - | 6 | 842 | 5389457 | ||
CX568915.1 | - | 5 | 260 | |||
D83073.1 | - | 6 | 697 | |||
U10871.1 | - | 6 | 764 |