Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172300.3 | NP_758504.3 | 7 | 1023 | ||
GenBank mRNA | AK005524.1 | - | 5 | 574 | ||
AK077793.1 | - | 7 | 1022 | |||
AK149770.1 | - | 7 | 1023 | |||
AK152529.1 | - | 7 | 737 | |||
AK153328.1 | - | 7 | 754 | |||
AK168942.1 | - | 7 | 716 | |||
AK187506.1 | - | 1 | 15 | |||
AK208933.1 | - | 3 | 142 | |||
BC027266.1 | - | 7 | 648 | |||
BC054412.1 | - | 7 | 726 | 2646768 |