Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138586.3 | NP_613052.1 | 4 | 485 | ||
GenBank mRNA | AK006475.1 | - | 4 | 438 | ||
AK007372.1 | - | 4 | 474 | |||
AK137780.1 | - | 448 | ||||
AK155228.1 | - | 4 | 437 | |||
AK189469.1 | - | 2 | 66 | |||
BC092393.1 | - | 2 | 2474 |