Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041075.1 | - | 4 | 443 | ||
BC125408.1 | - | 3 | 323 | |||
BC125410.1 | - | 3 | 323 |