Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB294520.1 | - | 5 | 498 | ||
AB294521.1 | - | 4 | 395 | |||
AB294522.1 | - | 4 | 387 | |||
AB294523.1 | - | 3 | 375 | |||
AB294524.1 | - | 3 | 339 | |||
AB294525.1 | - | 4 | 431 | |||
AB294526.1 | - | 4 | 395 | |||
AB294527.1 | - | 4 | 387 | |||
AB294528.1 | - | 3 | 339 | |||
AB300594.1 | - | 4 | 624 | |||
AK038726.1 | - | 4 | 682 | |||
AK053540.1 | - | 4 | 625 |