Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Endov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164636.1 | NP_001158108.1 | 6 | 600 | ||
NM_177394.3 | NP_796368.1 | 5 | 652 | |||
GenBank mRNA | AK042626.1 | - | 5 | 540 | ||
AK084233.1 | - | 6 | 602 | |||
AK084548.1 | - | 6 | 476 | |||
AK170595.1 | - | 5 | 645 | |||
AK172181.1 | - | 5 | 619 | |||
BC056486.1 | - | 5 | 572 | 5697870 |