Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mreg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005423.2 | NP_001005423.1 | 3 | 466 | ||
GenBank mRNA | AY628210.1 | - | 3 | 270 | ||
BC068125.1 | - | 3 | 466 | 6828150 |