Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205844.3 | NP_995316.2 | 6 | 1100 | ||
GenBank mRNA | AY457637.2 | - | 6 | 1100 |