Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028384.1 | NP_082660.1 | 4 | 451 | ||
GenBank mRNA | AK158659.1 | - | 2 | 231 | ||
BC147354.1 | - | 2 | 196 | 9056407 |