Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015821.2 | NP_056636.2 | 2 | 468 | ||
GenBank mRNA | AF176523.1 | - | 2 | 393 | ||
AK053206.1 | - | 2 | 434 | |||
AK155194.1 | - | 2 | 429 | |||
AK162522.1 | - | 1 | 682 | |||
AK170367.1 | - | 2 | 428 | |||
BC023902.1 | - | 1 | 333 | 5321302 | ||
CX222518.1 | - | 2 | 436 |