Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026631.3 | NP_080907.1 | 4 | 447 | ||
GenBank mRNA | AK007340.1 | - | 4 | 435 | ||
AK008766.1 | - | 4 | 474 | |||
AK019134.1 | - | 4 | 448 | |||
BC024944.1 | - | 4 | 438 | 3486985 |