Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145928.1 | NP_666040.1 | 6 | 534 | ||
GenBank mRNA | AK030593.1 | - | 6 | 536 | ||
AK156671.1 | - | 6 | 547 | |||
BC024611.1 | - | 6 | 539 | 4188261 | ||
BC025568.1 | - | 6 | 548 | 5322531 | ||
BC026574.1 | - | 6 | 534 | 4954874 |