Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018810.2 | NP_061280.2 | 5 | 1013 | ||
GenBank mRNA | AB026986.1 | - | 5 | 1118 | ||
AF192785.1 | - | 5 | 1038 | |||
AK005137.1 | - | 5 | 847 | |||
AK029500.1 | - | 5 | 1053 | |||
AK077060.1 | - | 5 | 1014 | |||
BC003329.1 | - | 5 | 1002 | 3158135 |