Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020033.1 | NP_064417.1 | 9 | 983 | ||
GenBank mRNA | AB076256.1 | - | 9 | 995 | ||
AJ011118.1 | - | 9 | 983 | |||
AJ245514.1 | - | 9 | 983 | |||
AK132585.1 | - | 9 | 993 | |||
BC138760.1 | - | 9 | 1021 | 8861782 | ||
BC138761.1 | - | 9 | 1021 | 8861783 |