Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122992.1 | NP_001116464.1 | 10 | 1041 | ||
NM_020273.2 | NP_064669.2 | 9 | 978 | |||
GenBank mRNA | AF210433.1 | - | 8 | 939 | ||
AK134349.1 | - | 9 | 979 | |||
BC046983.1 | - | 10 | 1012 | 6477130 | ||
CN459906.1 | - | 2 | 47 |